missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ SGE Liner for AC Control Injector
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092210
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent GC Systems
Specifications
Simulated Distillation Inlet Liner | |
73 mm | |
3 mm |
5/Pk. | |
1.6 mm | |
Programmed Injector |