Learn More
Trajan™ Glass Lined Bulkhead Unions with Locknut
Product Code. 15239163
Click to view available options
Diameter (Metric) Inner:
0.4 mm
0.5 mm
0.8 mm
Description:
0.25 mm restriction (at one end)
0.35 mm restriction (at one end)
No restriction
Unit Size:
Each
Product Code. 15239163
Brand: Trajan™ 1236306
Only null left
Add to basket
Add to basket
Glass lined stainless steel construcion. Bulkhead unions with locknut
  • Allows plate mounting of union
  • For use with fused silica
  • Low dead volume
  • High temperature applications
Specifications
Type Bulkhead Union
Description No restriction
Material Glass-lined Stainless Steel
Diameter (Metric) Inner 0.8 mm