Learn More
Trajan™ BP1 GC Capillary Columns
Product Code. 10045400
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Unit Size:
Each
Product Code. 10045400
Brand: Trajan™ 054047
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Max. Temperature 340/360°C
Functionality/Interaction Type Van der Waals interaction
Diameter (Metric) Inner 0.22 mm
Stationary Phase 100% Dimethyl Polysiloxane
USP Type G1, G2, G38
For Use With (Application) Hydrocarbon Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
Temperature Range -60°C to +340/360°C
Film Thickness 0.25 μm
Length (Metric) 25 m
Packing Material Forte™ BP1
Polarity Non polar
Coating Fused Silica