Learn More
SGE™ Beckman/Altex, Rheodyne, SSI, Valco Valves with Fixed Needle
Used with Rheodyne™ and Valco™ Valves. SGE Analytical Science™ Beckman/Altex, Rheodyne, SSI, Valco Valves with Fixed Needle have a 22 gauge.
Brand:  SGE™ 002315
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10005460
  • 271.00 EUR / Pack of 6
Estimated Shipment: 01-08-2024
Description
  • Fixed needle
  • For use with For Rheodyne™ and Valco™ Valves
Specifications
Specifications
Syringe
Beckman/Altex, Rheodyne™, SSI instruments, and valco valves
0.028 in.
10 μL
22 G
HPLC Valve Syringe with Fixed LC Needle
0.71 mm