Learn More
SGE™ Autosampler Syringes with 42mm Cone Tip Fixed Needle, 10 μL
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Pack of 6
Description
- Improved solvent resistance
- Wide temperature ranges
- Increased operational smoothness
- Color coded syringes by volume
- Range of needle gauge and length options
Specifications
Specifications
| Description | Not gas tight |
| Needle Gauge | 23 to 26 G |
| Diameter (Metric) Outer Needle | 0.63/0.47 mm |
| For Use With (Application) | Gas chromatography |
| Length (Metric) | 42 mm |
| Product Type | Autosampler Syringe |
| Model | 10F-AG-0.63/0.47C 6 pack |
| Volume (Metric) | 10 μL |
| Length (Metric) Needle | 42 mm |
| Needle Point Style | Cone |
| Show More |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering