missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETDB1 Monoclonal Antibody (5H6A12)

Description
MA5-15722 targets SETDB1 in indirect ELISA, IF and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15722 immunogen is purified recombinant fragment of human SETDB1 expressed in E. Coli. MA5-15722 detects SETDB1 which has a predicted molecular weight of approximately 180kDa.
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | SETDB1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5H6A12 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | SETDB1 |
| Gene Accession No. | O88974, Q15047 |
| Gene Alias | AU022152; ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; H3-K9-HMTase 4; H3-K9-HMTase4; histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; KG1T; KIAA0067; KMT1E; Lysine N-methyltransferase 1E; mKIAA0067; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; SETDB1; TDRD21; tudor domain containing 21 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?