missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB8 Monoclonal Antibody (anti-PI8K)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517647
This item is not returnable.
View return policy
Description
MA5-17647 targets PI-8/Proteinase Inhibitor 8 in ELISA and WB applications and shows reactivity with Human samples. The MA5-17647 immunogen is human Proteinase Inhibitor 8 (PI8) purified from the supernatant of thrombin activated platelets.
The superfamily of high molecular weight serine proteinase inhibitors. A critical portion of the molecule, the reactive center loop connects beta sheets A and C. Protease inhibitor-8 (PI8; SERPINB8) is a member of the ov-serpin subfamily, which, relative to the archetypal serpin PI1 (MIM 107400), is characterized by a high degree of homology to chicken ovalbumin, lack of N- and C-terminal extensions, absence of a signal peptide, and a serine rather than an asparagine residue at the penultimate position.
Specifications
| SERPINB8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P50452 | |
| SERPINB8 | |
| Human Proteinase Inhibitor 8 (PI8) purified from the supernatant of thrombin activated platelets. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| anti-PI8K | |
| Unconjugated | |
| SERPINB8 | |
| CAP2; CAP-2; Cytoplasmic antiproteinase 2; NK10; ovalbumin; peptidase inhibitor 8; PI8; PI-8; protease inhibitor 8 (ovalbumin type); serine (or cysteine) peptdiase inhibitor, clade B, member 8; serine (or cysteine) peptidase inhibitor, clade B, member 8; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 8; serine (or cysteine) proteinase inhibitor, clade B, member 8; serine protease inhibitor 8; serpin B8; serpin family B member 8; serpin peptidase inhibitor, clade B (ovalbumin), member 8; Serpinb8; Spi8 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction