missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A12, Mouse anti-Human, Clone: VP63, Novus Biologicals™
Mouse Monoclonal Antibody
380.00 EUR - 546.00 EUR
Specifications
| Antigen | Serpin A12 |
|---|---|
| Clone | VP63 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282506
|
Novus Biologicals
NBP2-80041-50UG |
50 μg |
380.00 EUR
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18274286
|
Novus Biologicals
NBP2-80041-100UG |
100 μg |
546.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A12 Monoclonal specifically detects Serpin A12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| Serpin A12 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| antitrypsin), member 12, member 12, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor | |
| SERPINA12 | |
| IgG1 κ | |
| Protein G purified | |
| Recognizes human Serpin A12. Detects a band of ∽48kDa by Western blot. |
| VP63 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| 145264 | |
| Recombinant human Serpin A12. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title