missing translation for 'onlineSavingsMsg'
Learn More

Serotonin, TRC

Product Code. 18030200
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030200 10 mg 10mg
18042520 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18030200 Supplier Toronto Research Chemicals Supplier No. TRCH97704310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-67-9
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.22
Synonym 1H-Indol-5-ol, 3-(2-aminoethyl)-, Indol-5-ol, 3-(2-aminoethyl)- (6CI,8CI), Tryptamine, 5-hydroxy-, 3-(2-Aminoethyl)-1H-indol-5-ol, 3-(2-Aminoethyl)indol-5-ol, 3-(β-Aminoethyl)-5-hydroxyindole, 5-HT, 5-HTA, 5-Hydroxy-3-(β-aminoethyl)indole, 5-Hydroxyltryptamine, 5-Hydroxytriptamine, 5-Hydroxytryptamine, DS substance, Enteramin, Enteramine, Serotonin, Substance DS, Thrombocytin, Thrombotonin
SMILES NCCc1c[nH]c2ccc(O)cc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H12N2O/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10/h1-2,5-6,12-13H,3-4,11H2
Formula Weight 176.095
Percent Purity >95%
Chemical Name or Material Serotonin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.