Learn More
Fisherbrand™ Septum for Magnetic Universal Precision Thread Cap
Shop All Fisherbrand Products
Click to view available options
Color:
Blue Transparent, White
Blue, White
Red, White
Silver, White
Material:
Aluminum, Silicone
PTFE, Silicone
Thickness (Metric):
1.3 mm
1.5 mm
Unit Size:
Pack of 1000
Specifications
Specifications
| Color | Silver, White |
| Diameter (Metric) Outer | 17 mm |
| Material | Aluminum, Silicone |
| Septum | Septa |
| Quantity | 1000 |
| Thickness (Metric) | 1.3 mm |