missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Septin 3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15769910 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15769910 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15769910 Supplier Invitrogen™ Supplier No. PA551959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LSKVVNIIPV IAKADTMTLE EKSEFKQRVR KELEVNGIEF YPQKEFDEDL EDKTENDKIR QESMPFAVVG SDKEYQVNGK RVLGRKTPWG IIEVENLNHC EFALLRDFVI RTHLQDLKEV THNIHYETYR AKRLNDNGG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

This gene belongs to the septin family of GTPases. Members of this family are required for cytokinesis. Expression is upregulated by retinoic acid in a human teratocarcinoma cell line. The specific function of this gene has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPT3
Gene Accession No. Q9UH03
Gene Alias 3110018K01Rik; 3-Sep; AV154067; B530002E20Rik; bK250D10.3; CTA-250D10.3; G-septin; G-septin alpha; MGC133218; neuronal-specific septin 3; neuronal-specific septin-3; OTTHUMP00000028742; P40; Sep3; SEPT3; septin 3; Septin3
Gene Symbols SEPT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Septin 3. Recombinant protein control fragment (Product #RP-91497).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55964
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.