Learn More
Macherey-Nagel™ Septa N 19 Polyethylene Caps
Product Code. p-8012410 Shop All Macherey-Nagel Products
Click to view available options
Color:
Beige, Gray
Red-Orange
Material:
Polyethylene, Butyl, PTFE
Polyethylene, Natural Rubber, TEF
Unit Size:
Pack of 100
Product Code. 10242983
Brand: Macherey-Nagel™ 70269
Only null left
Add to basket
Add to basket
Protect important material with the best septa. Macherey-Nagel™ Septa N 19 polyethylene caps for polyethelyne for N 20 caps.
  • Septa N 19 for PE caps N 20
  • Used as washer bottle closures or interim closures
  • Packs of 100 closures each
Specifications
Color Beige, Gray
Material Polyethylene, Butyl, PTFE
Septum Septa N 19
Quantity 100 Pk.
Thickness (Metric) 1.3 mm