Learn More
STEMCELL Technologies SepMate™-50 (IVD)
Description
- Simplify peripheral blood mononuclear cell (PBMC) isolation by incorporating SepMate into your density gradient centrifugation step
- SepMate tubes contain an insert that creates a barrier between the density gradient medium and blood, thus eliminating the need for careful blood layering and allowing mononuclear cells to be easily harvested with a simple pour
- This product can be used with RosetteSep to isolate specific immune cell subsets
- SepMate is manufactured under cGMP and registered as an In Vitro Diagnostic (IVD) device in Australia, Canada, Europe, and the USA
- In China, SepMate is considered a nonmedical device by the China Food and Drug Administration (CFDA) and should be used as general lab equipment
- The end user is responsible for determining whether the product is suitable for their specific application
- Users in all other countries, please refer to SepMate-15 (RUO) (Catalog #86415) and SepMate-50 (RUO) (Catalog #86450)
Specifications
Specifications
| For Use With | Cell Isolation; In Vitro Diagnostic; B Cells; Dendritic Cells; Monocytes; Mononuclear Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+; T Cells, Other Subsets; T Cells, Regulatory |
| Quantity | 500 Tubes |