Learn More
STEMCELL Technologies SepMate™-50 (IVD)
Product Code. 16506364
Encompass
Change view
Click to view available options
Quantity:
100 Tubes
500 Tubes
Unit Size:
Pack of 100
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16506364 500 Tubes Pack of 500
15654468 100 Tubes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16506364 Supplier STEMCELL Technologies Supplier No. 85460
50 mL centrifuge tube with an insert for density gradient centrifugation
  • Simplify peripheral blood mononuclear cell (PBMC) isolation by incorporating SepMate into your density gradient centrifugation step
  • SepMate tubes contain an insert that creates a barrier between the density gradient medium and blood, thus eliminating the need for careful blood layering and allowing mononuclear cells to be easily harvested with a simple pour
  • This product can be used with RosetteSep to isolate specific immune cell subsets
  • SepMate is manufactured under cGMP and registered as an In Vitro Diagnostic (IVD) device in Australia, Canada, Europe, and the USA
  • In China, SepMate is considered a nonmedical device by the China Food and Drug Administration (CFDA) and should be used as general lab equipment
  • The end user is responsible for determining whether the product is suitable for their specific application
  • Users in all other countries, please refer to SepMate-15 (RUO) (Catalog #86415) and SepMate-50 (RUO) (Catalog #86450)
Specifications
For Use With Cell Isolation; In Vitro Diagnostic; B Cells; Dendritic Cells; Monocytes; Mononuclear Cells; NK Cells; T Cells; T Cells, CD4+; T Cells, CD8+; T Cells, Other Subsets; T Cells, Regulatory
Quantity 500 Tubes