Learn More
WTW™ SenTix™ 940 Digital IDS pH/ORP Electrodes
GREENER_CHOICE
Product Code. 11705174
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
Product Code. 11705174
Brand: WTW™ 103740
Only null left
Add to basket
Add to basket
Provides optimum digital measurement of pH with built-in electrode quality monitoring that is usable in all areas of lab and field measurement.
  • Low-maintenance IDS pH gel electrode, 1,5 m fixed cable
  • Digital measurement of pH with built-in electrode quality monitoring - usable in all areas of lab and field measurement
  • Safely measured values due to signal processing in the sensor
  • Easy electrode replacement thanks to calibration data stored inside sensor
  • Sensor quality monitoring by QSC function
  • IDS pH electrodes for field use, all the way to special applications in the lab
  • QSC function allows monitoring of the current sensor status with all IDS pH electrodes
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Fixed Cable
Junction Fiber
Material Plastic Shaft, Fiber Diaphragm
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type IDS pH/ORP Electrode
Includes 1.5 m Cable
Model SenTix™ 940
pH Relative Accuracy ±0.004, ±1
Probe NTC
Storage Requirements Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature Accuracy ±0.2°C
Temperature (Metric) Range 0°C to 80°C
Length (Metric) Cable 1.5 m
Length (Metric) Stem 120 mm