Learn More
Cytiva Sentino™ GN-6 Membrane
Click to view available options
:
0.45 μm
Description
- Ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
- Gamma irradiated and ready to use
- For research use only
Specifications
Specifications
| Material (Membrane) | MCE |
| Pore Size | 0.45 μm |
| Sterility | Sterile |
| Includes | Black Grid Lines |
| Type | Sentino Dispenser Pack |