Learn More
Ansell™ Sensilite™ 48-135 Series White and Gray Polyurethane Light Weight Gloves
Product Code. 15240009 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15240009 8 12 pairs
15260009 10 12 pairs
15220009 6 12 pairs
15270009 11 12 pairs
15250009 9 12 pairs
15230009 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15240009 Supplier Ansell™ Supplier No. 48135080
Only null left
Add to basket
Add to basket
Meets most electrostatic discharge protection needs and provides flexibility and dexterity.
  • Special purpose gloves
  • 15-gauge nylon liner
  • Added carbon fibre offers economical product-protection solution
  • Comfort and good fit are assured through seamless construction and a wide range of sizes
  • With the palm dipped in a polyurethane coating, offers good mechanical resistance and durability
  • Fingertip-dipped version allows better fit and flexibility
  • Coated fingertip also prevents the transfer of perspiration to the electronic part
Applications:
  • Final assembly of dashboard, cables and wires
  • Final assembly of electronic components and mother boards
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 113X
Cuff Material Polyurethane
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyurethane
Product Type Lightweight Gloves
Size 8
Finger Style Coated-fingertip
Glove Cuff Style Knit
Liner Material Nylon Carbon Fiber
Lining Lined
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Good mechanical resistance
Series 48-135