Learn More
Ansell Edmont™ Sensilite™ 48-120 Series White Polyurethane Light Weight Gloves
Product Code. 12690932 Shop All Ansell Products
Change view
Click to view available options
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
12690932 12 pairs
12650932 12 pairs
12660932 12 pairs
12640932 12 pairs
12670932 12 pairs
12680932 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12690932 Supplier Ansell Edmont™ Supplier No. 48120090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Economical hand protection solution offering good grip, dexterity and comfort.
  • Multi-purpose gloves
  • Polyester liner offers good hand protection in lightweight applications
  • Polyurethane coating on the palm resists tears and abrasions
Applications:
  • Assembly of small pieces such as small bearings, joints and springs
  • Quality control
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 3121
Cuff Material Polyurethane
Coating Material Polyurethane
Product Type Lightweight Gloves
Style 48-120
Glove Cuff Style Knitwrist
Lining Polyester
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Tears and abrasions
Series 48-120