Learn More
Ansell Edmont™ Sensilite™ 48-120 Series White Polyurethane Light Weight Gloves
Shop All Ansell Products
Click to view available options
Unit Size:
12 pairs
Description
- Multi-purpose gloves
- Polyester liner offers good hand protection in lightweight applications
- Polyurethane coating on the palm resists tears and abrasions
Applications:
- Assembly of small pieces such as small bearings, joints and springs
- Quality control
Specifications
Specifications
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, EN 388 : 3121 |
| Cuff Material | Polyurethane |
| Coating Material | Polyurethane |
| Product Type | Lightweight Gloves |
| Style | 48-120 |
| Glove Cuff Style | Knitwrist |
| Lining | Polyester |
| Palm Type | Coated-palm |
| Quantity | 144 Pairs |
| Show More |