missing translation for 'onlineSavingsMsg'
Learn More

Semaxinib, MedChemExpress

Product Code. p-200096116
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16480267

missing translation for 'mfr': Medchem Express HY10374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Semaxinib (SU5416) is a potent and selective inhibitor of VEGFR (Flk-1/KDR) with an IC50 of 1.23 μM.

Semaxinib (SU5416) is a potent and selective inhibitor of VEGFR (Flk-1/KDR) with an IC50 of 1.23 μM.

Specifications

Chemical Name or Material Semaxinib
Physical Form Solid
CAS 204005-46-9
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H14N2O
For Use With (Application) Cancer-Kinase/protease
Synonym SU5416
Solubility Information DMF : 50 mg/mL (209.84 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (83.93 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC=C/21)C2=C/C3=C(C)C=C(C)N3
Molecular Weight (g/mol) 238.28
Formula Weight 238.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.