Learn More
Semaxinib, MedChemExpress
Product Code. 16480267
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16480267
Brand: Medchem Express HY10374
MedChemExpress Semaxinib (SU5416) is a potent and selective inhibitor of VEGFR (Flk-1/KDR) with an IC50 of 1.23 μM.
Semaxinib (SU5416) is a potent and selective inhibitor of VEGFR (Flk-1/KDR) with an IC50 of 1.23 μM.
Specifications
Chemical Name or Material Semaxinib
Physical Form Solid
CAS 204005-46-9
Color Orange
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H14N2O
For Use With (Application) Cancer-Kinase/protease
Synonym SU5416
Solubility Information DMF : 50 mg/mL (209.84 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (83.93 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC=C/21)C2=C/C3=C(C)C=C(C)N3
Molecular Weight (g/mol) 238.28
Formula Weight 238.28
Percent Purity 98.0%
Grade Research