missing translation for 'onlineSavingsMsg'
Learn More

Selonsertib, MedChemExpress

Product Code. p-200100544
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483657

Brand: Medchem Express HY18938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Selonsertib (GS-4997), an orally bioavailable, selective apoptosis signal-regulating kinase 1 (ASK1) inhibitor with a pIC50 of 8.3, has been evaluated as an experimental treatment for diabetic nephropathy and kidney fibrosis.

Selonsertib (GS-4997), an orally bioavailable, selective apoptosis signal-regulating kinase 1 (ASK1) inhibitor with a pIC50 of 8.3, has been evaluated as an experimental treatment for diabetic nephropathy and kidney fibrosis.

Specifications

Chemical Name or Material Selonsertib
CAS 1448428-04-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H24FN7O
Quantity 50 mg
Synonym GS-4997
Solubility Information DMSO : ≥ 31 mg/mL (69.59 mM)
SMILES O=C(NC1=NC(C2=NN=CN2C(C)C)=CC=C1)C3=CC(N4C=C(C5CC5)N=C4)=C(C)C=C3F
Molecular Weight (g/mol) 445.49
Formula Weight 445.49
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.