missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seladelpar sodium salt, MedChemExpress
Description
Seladelpar sodium salt (MBX-8025) is an orally active, potent and specific PPARδ agonist with an EC50 of 2 nM, showing more than 750-fold and 2500-fold selectivity over the PPARα and PPARγ receptors, respectively.
Specifications
Specifications
| Chemical Name or Material | Seladelpar sodium salt |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H22F3NaO5S |
| Quantity | 10 mg |
| Synonym | MBX-8025 sodium salt RWJ-800025 sodium salt |
| Solubility Information | DMSO : 50 mg/mL (107.19 mM; Need ultrasonic) |
| SMILES | O=C(O[Na])COC1=CC=C(SC[C@H](OCC)COC2=CC=C(C(F)(F)F)C=C2)C=C1C |
| Molecular Weight (g/mol) | 466.45 |
| Formula Weight | 466.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?