Learn More
Kinematica™ Security Lid for Mixing Attachment
Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
MBA 1000 Mixing Attachment
MBA 250 Mixing Attachment
MBA 500 Mixing Attachment
Height (Metric):
128 mm
149 mm
228 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Security Lid |
| Height (Metric) | 128 mm |
| Material | Protection Glass |
| For Use With (Equipment) | MBA 250 Mixing Attachment |