Learn More
BINDER™ Rack Fastener
Shop All BINDER Products
missing translation for 'orderingAttributeHoverText'
Material:
Steel
missing translation for 'unitSize'
Each
Description
- One set of four fasteners for additional security
- Securing element for racks
Specifications
Specifications
| Description | Rack accessories - fasteners (1 set of 4) for additional security of racks |
| For Use With (Equipment) | KB 23, KB 53, KB 115, FD 23, FED 400, FP 53, FP 115, FP 240, FP 400, FP 720, M 53, M115, M 240, M 400, M 720 |
| Material | Steel |
| Type | Rack Fastener |
| Includes | 1 Set of four fasteners |