Learn More
sec-Butyl acetate, 98%
Product Code. 11383297
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383297 25 g 25g
11393297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11383297 Supplier Thermo Scientific Alfa Aesar Supplier No. L13892.14
Request Bulk or Custom Format
CAS: 105-46-4 | C6H12O2 | 116.16 g/mol
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105-46-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009328
InChI Key DCKVNWZUADLDEH-UHFFFAOYNA-N
Synonym sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
PubChem CID 7758
IUPAC Name butan-2-yl acetate
SMILES CCC(C)OC(C)=O
Specifications
Melting Point -99°C
Density 0.872
Boiling Point 111°C to 112°C
Flash Point 16°C (60°F)
Refractive Index 1.389
Quantity 25 g
UN Number UN1123
Beilstein 1720689
Merck Index 14,1536
Solubility Information Slightly soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material sec-Butyl acetate