Learn More
Lonza™ SeaKem™ LE Agarose
Product Code. 11510727
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11510727 1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11510727 Supplier Lonza™ Supplier No. 50005
For analysis of nucleic acids by gel electrophoresis and blotting. Lonza SeaKem™ LE Agarose is recommended for protein gel electrophoresis in applications such as ouchterlony and radial immunodiffusion (RID).
  • Each gel sharply resolves DNA and provides consistent resolution from lot-to-lot
  • Agarose has no detectable DNase or RNase activity
  • Forms strong gels with low background upon ethidium bromide, SYBR™ Green, or GelStar™ staining
  • Certificate of analysis is provided
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Ouchterlony and radial immunodiffusion (RID)
Quantity 1 kg
Product Type Agarose Gel Cassette