Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spacers
Product Code. 10668285 Shop All Cytiva Products
Change view
Click to view available options
Thickness (Metric):
0.1 cm
0.15 cm
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) unitSize
10668285 0.1 cm Pack of 2
10065794 0.15 cm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10668285 Supplier Cytiva Supplier No. SE2619T21.0
Only null left
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type T-shape Spacer
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Length (Metric) 1.05 cm