Learn More
Kimberly-Clark™ Scott™ Performance Standard Toilet Tissue Rolls
Click to view available options
Packaging Quantity:
6 x 6 Rolls x 350 Sheets
8 x 8 Rolls x 350 Sheets
Unit Size:
64 rolls
Pack of 36
Description
- White, perforated, 2-ply toilet tissue
- Soft and absorbent
- Dissolves in cold water so that the sewer pipes will not clog up
- Perforation allows economical consumption, suitable for cost savings and reducing the amount of waste
- ISO 14001, ISO 9001, EU Label and FSC certified
- Ideal for washrooms in office buildings and hotels that value economy
Specifications
Specifications
| Product Type | Toilet Tissue Roll |
| Length (English) | 4.72 in. |
| Length (Metric) | 12 cm |
| Width (English) | 3.7 in. |
| Width (Metric) | 9.3 cm |
| Material | Paper |
| Certifications/Compliance | ISO 14001, ISO 9001, EU Ecolabel and FSC certified |
| Color | White |
| Dimensions (L x W) | 12 x 9.3 cm (4.72 x 3.66 in.) |
| For Use With (Application) | Ideal for small washrooms in office buildings and hotels, the place to economy value |
| Show More |