missing translation for 'onlineSavingsMsg'
Learn More

Scopine Hydrochloride, TRC

Product Code. 30414155
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414155 50 mg
30423972 250 mg
30423866 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414155 Supplier Toronto Research Chemicals Supplier No. TRCS19997550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85700-55-6
Molecular Formula C8 H13 N O2 . Cl H
Molecular Weight (g/mol) 191.66
Synonym 3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 9-methyl-, hydrochloride (1:1), (1α,2β,4β,5α,7β)-, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol, 9-methyl-, hydrochloride, (1α,2β,4β,5α,7β)- (9CI), Scopine hydrochloride
IUPAC Name (1S,2S,4R,5R)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-ol;hydrochloride
SMILES Cl.CN1[C@@H]2C[C@@H](O)C[C@H]1[C@@H]3O[C@H]23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H13NO2.ClH/c1-9-5-2-4(10)3-6(9)8-7(5)11-8;/h4-8,10H,2-3H2,1H3;1H/t4-,5-,6+,7-,8+;
Formula Weight 191.0713
Chemical Name or Material Scopine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.