Description
- ScienceLine micro electrode
- Spear tip
- Surface pH electrode
- Pt 1000 sensor
Specifications
Specifications
| Connector Types | BNC |
| Diameter (Metric) Body | 12/5 mm |
| Length (Metric) | 40/130 mm |
| Material | Platinum |
| Model | A 157 1M-BNC-ID |
| Temperature (Metric) Range | -5°C to 100°C |
| Includes | Sensor Cognition |
| Type | Combination Electrode |