Learn More
Cole-Parmer™ Schott™ FEP Tubing
Shop All Cole Parmer Products
Click to view available options
Overall Length (English):
40 in.
Unit Size:
Each
Description
- For use with SCHOTT Volumetric Titrator, 24906-00.
Specifications
Specifications
| Material | FEP |
| Overall Length (English) | 40 in. |
| Overall Length (Metric) | 101.6 cm |
| For Use With | Schott™ Volumetric Titrator, 24906-00 |
| Product Type | Tubing for Titrator 24906-00 |