missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) acetate hydrate, 99.9% (metals basis)

Product Code. p-7024084
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11463817

Brand: Thermo Scientific Alfa Aesar A18271.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 304675-64-7 | C6H9O6Sc | 222.09 g/mol

Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304675-64-7
Molecular Formula C6H9O6Sc
Molecular Weight (g/mol) 222.09
MDL Number MFCD00150122
InChI Key DBTMQFKUVICLQN-UHFFFAOYSA-K
IUPAC Name scandium(3+) triacetate
SMILES [Sc+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifikationer

Linear Formula (CH3CO2)3Sc·xH2O
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 222.10 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Scandium(III) acetate hydrate

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.