Learn More
SB-772077B dihydrochloride, MedChemExpress
Product Code. 16486917
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486917 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16486917 Supplier Medchem Express Supplier No. HY108518
MedChemExpress SB-772077B dihydrochloride is an aminofurazan-based Rho kinase (ROCK) inhibitor with IC50s of 5.6 nM and 6 nM toward ROCK1 and ROCK2, respevtively.
SB-772077B dihydrochloride is an aminofurazan-based Rho kinase (ROCK) inhibitor with IC50s of 5.6 nM and 6 nM toward ROCK1 and ROCK2, respevtively.
Specifications
Chemical Name or Material SB-772077B dihydrochloride
CAS 607373-46-6
Color White
Purity Grade Notes Research
Molecular Formula C15H20Cl2N8O2
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (240.80 mM; Need ultrasonic) ∣DMSO : 38.33 mg/mL (92.30 mM; ultrasonic and warming and heat to 80°C)
SMILES [H]Cl.O=C(C1=CN=CC2=C1N(CC)C(C3=NON=C3N)=N2)N4C[C@@H](N)CC4.[H]Cl
Molecular Weight (g/mol) 415.28
Formula Weight 415.28
Percent Purity 98.04%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation