Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
Product Code. 11708569 Shop All Thermo Scientific Products
Click to view available options
Holds:
1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
12 Deep-well Plates
192 Vials
30 x 15 mL Conical Centrifuge Tubes
52 x 15 mL Conical
8 Microwell Plates
Flasks
Unit Size:
Each
Pack of 2
Product Code. 11708569
Brand: Thermo Scientific™ MPTR12210
Only null left
Add to basket
Add to basket
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications
Description Microwell Plates: 12 Shallow-well, Large capacity models
Type Rotor
For Use With (Equipment) SPD210, SPD2030, SPD300DDA (requires RAD200)
Holds 12 Deep-well Plates
Model MPTR12-210
Rotor System Traditional