Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Shop All Thermo Scientific Products
Click to view available options
Description:
SpeedVac SPD300DDA Vacuum Concentrator Kit
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor
SpeedVac SPD300DDA Vacuum concentrator
Electrical Requirements:
230 V 50 Hz
Unit Size:
Each
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
- For strong acids and bases used in combinatorial chemistry
Specifications
Specifications
| Certifications | UL Listed to US and Canadian requirements. CE marked |
| Description | SpeedVac SPD300DDA Vacuum concentrator |
| Chamber Material | Aluminum |
| Model | SPD300 |
| Temperature Range (Metric) | 35°C to 85°C |
| Electrical Requirements | 230 V 50 Hz |
Product Content Correction
Product Title
Spot an opportunity for improvement?Übermitteln Sie eine inhaltliche Korrektur