Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Shop All Thermo Scientific Products
Click to view available options
Holds:
10 Centrifuge Tubes
100 (6 x 50mm) Tubes
100 (8 x 29 mm) Tubes
12 Scintillation Vials
12 Vials
24 Vials
32 Tubes
4 x 100 mL Pear-shaped Flasks
40 Microtubes
40 Tubes
6 x 50 mL Conical Tubes
60 Vials
64 Microtubes
72 Tubes
8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
Unit Size:
Each
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
- Easy to use
- Multi-size tube holes accommodate different sample sizes
- Require no additional accessories
- Fit directly into your concentrator
Specifications
Specifications
| Description | Microtubes: 64 x 1.5mL, Medium capacity models |
| Type | Rotor |
| For Use With (Equipment) | SPD1030, SPD120, SPD130, SPD140DDA |
| Holds | 64 Microtubes |
| Model | RH64-11 |
| Rotor System | Traditional |
| Volume (Metric) | 1.5 mL |