Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Shop All Thermo Scientific Products
Click to view available options
Type:
Acid Neutralization Cartridge
Acid Neutralizing Compound
Ammonia Neutralizing Solution
Cart
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Unit Size:
0.25kg
5 L
Each
Pack of 4
Description
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Specifications
Specifications
| Description | Ammonia Neutralizing Solution (Oligo Prep) |
| Type | Ammonia Neutralizing Solution |
| For Use With (Equipment) | All SpeedVac™ Unit |
| Model | ANS121 |
| Quantity | Each |
Safety and Handling
Signal Word
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337-If eye irritation persists:.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.