Learn More
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Shop All Thermo Scientific Products
Click to view available options
Type:
Acid Neutralization Cartridge
Acid Neutralizing Compound
Ammonia Neutralizing Solution
Cart
Chemical Trap Kit
Condensation Flask
Disposable Cartridge
Heat Transfer Fluid
Unit Size:
0.25kg
5 L
Each
Pack of 4
Description
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Specifications
Specifications
| Description | Disposable Cartridge (acid neutralization) |
| Type | Acid Neutralization Cartridge |
| For Use With (Equipment) | All SpeedVac™ Unit |
| Model | DC120A |