Learn More
Sartorius Vivaflow™ Models Accessories: Female Luer Fittings
Shop All Sartorius Products
Click to view available options
Type:
Size 15 Female Luer Fitting
Size 16 Female Luer Fitting
Unit Size:
Pack of 10
Description
- Manage and maintain flow
Specifications
Specifications
| Connectors | Female Luer Size 16 |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Vivaflow 50 and 200 |
| Material | PVC |
| Sterility | Non-sterile |
| Quantity | 10/Pk. |
| Type | Size 16 Female Luer Fitting |