Learn More
Sartorius Vivaflow™ Models Accessories: Diafiltration Reservoir
Shop All Sartorius Products
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Each
Description
- Manage and maintain flow
Specifications
Specifications
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Vivaflow 50 and 200 |
| Sample Volume | 500 mL |
| Sterility | Non-sterile |
| Quantity | Each |
| Type | Diafiltration Reservoir |
| Volume (Metric) | 500 mL |