Learn More
Sartorius Disposable Plastic Syringes
Shop All Sartorius Products
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
50 mL
Unit Size:
Pack of 12
Description
- Male luer-lok connector
- Comes with needle
- Sterile
Specifications
Specifications
| Material | Plastic |
| Needle Gauge | 19 G |
| For Use With (Application) | For filter holders |
| Product Type | Syringe |
| Disposable | Single-use |
| Volume (Metric) | 10 mL |
| Needle Point Style | Cone |
| Connection Type | Male Luer Lock |
| Sterility | Sterile |