Learn More
Sartorius arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)
BIOPHARMA
Product Code. p-4597934 Shop All Sartorius Products
Change view
Click to view available options
Mounting:
Benchtop
Wall
Includes:
TOC Monitor
UV Lamp, TOC Monitor
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Includes unitSize
13497058 Benchtop TOC Monitor Each
15164409 Wall TOC Monitor Each
17119084 Benchtop UV Lamp, TOC Monitor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13497058 Supplier Sartorius Supplier No. H2OI2TOCT
Use the Sartorius arium™ Comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
  • Reliable TOC content of <2 ppb for reproducible results
  • Automatically optimised water consumption with iJust
  • Easy-to-use - unique glass display with touch function
Benefits:
  • Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
  • Optimized water consumption
  • Meet customer's lab requirements and saves maintenance time
  • Safe operation
  • Reliable TOC reduction
  • Supports reliable recording of important data
  • Easy operation
  • HPLC
  • GC-MS, AAS, ICP-MS
  • Ion chromatography
  • TOC-Analysis
  • Photometry
  • Buffer and pH-solutions
  • Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
TRUSTED_SUSTAINABILITY
Specifications
Conductivity 0.055 Product water, Less than1500 feed water
Contaminant Rejection (Microbiological) Greater than99%
Contaminant Rejection (Organic) Greater than99%
Bacterial Content <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Feedwater Connector 3/8 in.
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate Up to 2 L/min.
Product Water Type Type 1 to 3
Remote Dispenser No
Resistivity 18.2 Mohm.cm
Total Organic Carbon ≤2 ppb
Feedwater Temperature (English) 35.6°F to 86°F
Feedwater Temperature (Metric) 2°C to 30°C
For Use With (Application) Feed water for ultrapure water system, rinsing Glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
Humidity Range 80%
Includes TOC Monitor
Mounting Benchtop
Particle Size <1/mL
pH Range 4 to 10
Purification Method Adsorption by spherical activated Carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration
Storage Requirements 5°C to 45°C
Temperature Range (English) 35.6°F to 95°F
Temperature Range (Metric) 2°C to 35°C
Tolerance <4
Turbidity <1
Width (English) 17.12 in.
Width (Metric) 43.5 cm
Height (English) 19.7 in.
Height (Metric) 50.1 cm
Depth (English) 18.74 in.
Depth (Metric) 47.6 cm
Weight (English) 68.3 lb.
Weight (Metric) 31 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Daily Output 120 L
Endotoxin Concentration <0.001 EU/mL with a terminal ultrafiltration cartridge
Product Type Arium Comfort I Combined System