missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF8 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18743527
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18743527 100 μg 100µg
18741897 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18743527 Supplier R&D Systems Supplier No. MAB10820100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ORF8 Monoclonal antibody specifically detects ORF8 in SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORF8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1041422
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_009724369
Host Species Mouse
Immunogen E. coli-derived human SARS-CoV-2 ORF8, Met16-His121, Accession # YP_009724369
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species SARS-CoV-2
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.