missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 Nucleocapsid Protein Titer Assay Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
Specifications
Specifications
| Product Type | Antigen assay kits |
| Kit Contents | This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Nucleocapsid Protein. The kit consists of Anti-SARS-CoV-2 Nucleocapsid Antibody, an SARS-CoV-2 Nucleocapsid Protein as Control, an biotin-Anti-SARS-CoV-2 N |
| Quantity | 96 Tests |
| Shipping Condition | RT |
| Storage Requirements | -20°C, aviod light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction