missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Phosphoprotein Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 EUR - 349.00 EUR
Specifications
| Antigen | SARS-CoV-2 Nucleocapsid Phosphoprotein |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233206
|
Proteintech
28769-1-AP-20UL |
20 μL |
145.00 EUR 129.00 EUR / 20µL Save 16.00 EUR 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236994
|
Proteintech
28769-1-AP-150UL |
150 μL |
410.00 EUR 349.00 EUR / 150µL Save 61.00 EUR 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of virus SARS-CoV-2 Nucleocapsid Phosphoprotein. This antibody recognizes virus antigen. The SARS-CoV-2 Nucleocapsid Phosphoprotein antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SARS-CoV-2 Nucleocapsid Phosphoprotein | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 43740575 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title