missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAR1B Polyclonal Antibody

Product Code. 15759531 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15759531 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15759531 Supplier Invitrogen™ Supplier No. PA561724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TLLHMLKDDR LGQHVPTLHP TSEELTIAGM TFTTFDLGGH VQARRVWKNY LPAINGIVFL VDCADHE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

The protein encoded by this gene is a small GTPase that acts as a homodimer. The encoded protein is activated by the guanine nucleotide exchange factor PREB and is involved in protein transport from the endoplasmic reticulum to the Golgi. This protein is part of the COPII coat complex. Defects in this gene are a cause of chylomicron retention disease (CMRD), also known as Anderson disease (ANDD). Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAR1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAR1B
Gene Accession No. Q9Y6B6
Gene Alias 2310075M17Rik; 2900019I22Rik; ANDD; CMRD; GTBPB; GTP-binding protein B; GTP-binding protein SAR1b; GTP-binding protein Sara; SAR1 gene homolog B; SAR1 gene homolog B (S. cerevisiae); SAR1 homolog B; SAR1a gene homolog 2; Sar1b; SAR1b gene homolog; SARA1B; SARA2; SARB; secretion associated Ras related GTPase 1B; secretion associated, Ras related GTPase 1B
Gene Symbols SAR1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAR1B. Recombinant protein control fragment (Product #RP-100036).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.