Learn More
Skytec™ Sapphire Total Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
missing translation for 'unitSize'
10 pairs
Description
- Oil Resistant Water Repellent Cut Resistant Grip Glove
- Cut resistant: Level E/ ANSI A5
- A flexible
- No irritation by seams in contact with the skin
- Elasticated wrist provides a secure fit
Specifications
Specifications
| Certifications/Compliance (Europe) | CE II, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 4X43E, EN 407:2020 X1XXXX, EN 21420:2020 |
| Construction | Seamless Knit |
| Coating Material | Foam Nitrile |
| Color | Black |
| Cut Resistance Rating | Level E |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| Show More |