Learn More
Masterflex™ Sanitary PVDF Clamps
Product Code. 11709589
Change view
Click to view available options
Size (Metric) Grip:
25.4 to 38.1 mm
50.8 mm
63.5 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Grip unitSize
11709589 25.4 to 38.1 mm Each
11719589 50.8 mm Each
15352830 63.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11709589 Supplier Masterflex™ Supplier No. 3181101
Only null left
Specifications
Autoclavable Autoclavable
Quantity 1 Ea.
Type Clamp
Material PVDF
For Use With (Equipment) Sanitary Fitting
Size (Metric) Grip 25.4 to 38.1 mm
Temperature (Metric) Resistant 138°C