Learn More
Sampling Systems™ Sampler Rack
Product Code. 15809571 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
500 mm
900 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15809571 500 mm Each
15899561 900 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15809571 Supplier Sampling Systems™ Supplier No. 8820A02
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These Sampler Racks are ideal for hanging up samplers when they are not in use.
  • Stainless steel and plastic construction
  • Standard rack can hold up to 5 samplers
  • Samplers are attached by a simple push clip design
  • Comes complete with hole fixings
  • Suitable for samplers from 10 to 35mm in diameter and up to 4kg in weight
Specifications
Product Type Storage Rack
Material Stainless Steel, Plastic
For Use With (Application) Sampler storage
Length (Metric) 500 mm