Learn More
Buerkle™ Stainless Steel Sample Spoon
Shop All Bürkle Products
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
300 mm
400 mm
500 mm
Unit Size:
Each
Description
- Made of AISI 304 (1.4301) stainless steel
- Largely chemical resistant (not against acids and alkalis)
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Spoon |
| For Use With (Application) | For Withdrawing Small Quantities of Samples, Foodstuffs |
| Material | Stainless Steel (V2A) |
| Length (Metric) | 400 mm |