Learn More
RETSCH Sample Splitters
Product Code. p-8007377 Shop All Retsch Products
Change view
Click to view available options
Depth (Metric):
6.3 mm
12.5 mm
25 mm
37.5 mm
50 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
10572121 50 mm Each
10562121 25 mm Each
10379870 6.3 mm Each
10432691 12.5 mm Each
10339780 37.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10572121 Supplier RETSCH Supplier No. 40.610.0005
Only null left
Specifications
Product Type Sample Splitter
Depth (Metric) 50 mm
Includes 3 Collecting Receptacles 8L Each, Stand and Dividing Head, all Made of Galvanized Sheet
Particle Size 50 mm